Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lect1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010701.2 | NP_034831.2 | 3 | 499 | ||
GenBank mRNA | AK013975.1 | - | 3 | 413 | ||
BC045152.1 | - | 3 | 398 | 6439864 | ||
U43509.1 | - | 3 | 500 |