Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lepr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122899.1 | NP_001116371.1 | 16 | 2963 | ||
NM_010704.2 | NP_034834.1 | 16 | 2963 | |||
NM_146146.2 | NP_666258.2 | 16 | 2963 | |||
GenBank mRNA | AK136231.1 | - | 2 | 696 | ||
AK140785.1 | - | 16 | 2465 | |||
AK143989.1 | - | 16 | 2652 | |||
AK155891.1 | - | 16 | 2504 | |||
BC082551.1 | - | 16 | 2220 | 30621706 | ||
U42467.1 | - | 16 | 2471 | |||
U46135.1 | - | 16 | 2471 | |||
U49106.1 | - | 15 | 2411 | |||
U49107.1 | - | 3 | 424 | |||
U49108.1 | - | 3 | 424 | |||
U49109.1 | - | 15 | 2411 | |||
U52915.1 | - | 16 | 2925 | |||
U58861.1 | - | 16 | 2493 | |||
U58862.1 | - | 16 | 2503 | |||
U58863.1 | - | 16 | 2493 | |||
Y10296.1 | - | 15 | 2419 | |||
Y10298.1 | - | 15 | 2419 |