Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lepr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122899.1 | NP_001116371.1 | 16, 17 | 3040 | ||
NM_010704.2 | NP_034834.1 | 16, 17 | 3040 | |||
NM_146146.2 | NP_666258.2 | 16, 17 | 3040 | |||
GenBank mRNA | AK136231.1 | - | 2, 3 | 773 | ||
AK140785.1 | - | 16, 17 | 2542 | |||
AK143989.1 | - | 16, 17 | 2729 | |||
AK155891.1 | - | 16, 17 | 2581 | |||
BC082551.1 | - | 16, 17 | 2297 | 30621706 | ||
U42467.1 | - | 16, 17 | 2548 | |||
U46135.1 | - | 16, 17 | 2548 | |||
U49106.1 | - | 15, 16 | 2488 | |||
U49107.1 | - | 3, 4 | 501 | |||
U49108.1 | - | 3, 4 | 501 | |||
U49109.1 | - | 15, 16 | 2488 | |||
U52915.1 | - | 16, 17 | 3002 | |||
U58861.1 | - | 16, 17 | 2570 | |||
U58862.1 | - | 16, 17 | 2580 | |||
U58863.1 | - | 16, 17 | 2570 | |||
Y10296.1 | - | 15, 16 | 2496 | |||
Y10298.1 | - | 15, 16 | 2496 |