Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008494.3 | NP_032520.1 | 3 | 624 | ||
GenBank mRNA | AF015768.1 | - | 3 | 653 | ||
AK004642.1 | - | 3 | 625 | |||
AK153283.1 | - | 3 | 625 | |||
BC115680.1 | - | 2 | 290 | 40104178 | ||
BC138600.1 | - | 3 | 645 | 8861620 | ||
BC138601.1 | - | 3 | 645 | 8861621 | ||
U94351.1 | - | 3 | 589 |