Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159301.1 | NP_001152773.1 | 3 | 319 | ||
NM_010708.2 | NP_034838.2 | 3 | 319 | |||
GenBank mRNA | AK146044.1 | - | 3 | 275 | ||
BC003754.1 | - | 3 | 246 | 3601419 | ||
CA458877.1 | - | 3 | 309 | |||
CA546274.1 | - | 3 | 319 | |||
U55060.1 | - | 3 | 245 | |||
U55061.1 | - | 3 | 226 |