Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008498.2 | NP_032524.1 | 3 | 1877 | ||
GenBank mRNA | AK032067.1 | - | 68 | |||
AK141086.1 | - | 1879 | ||||
BC092374.1 | - | 3 | 2010 | 30363447 | ||
S68107.1 | - | 6 | 1409 | |||
Z27410.1 | - | 3 | 1764 |