Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039653.2 | NP_001034742.1 | 5 | 837 | ||
NM_010711.2 | NP_034841.2 | 5 | 754 | |||
GenBank mRNA | AK047273.1 | - | 2 | 597 | ||
BC132554.1 | - | 5 | 751 | 40130831 | ||
BC132556.1 | - | 5 | 751 | 40130833 | ||
BC150689.1 | - | 5 | 723 | 9087600 | ||
L38248.1 | - | 5 | 803 | |||
L38857.1 | - | 5 | 754 |