Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291245.1 | NP_001278174.1 | 13, 14 | 2162 | ||
NM_001291246.1 | NP_001278175.1 | 13, 14 | 2159 | |||
NM_001291247.1 | NP_001278176.1 | 13, 14 | 2163 | |||
NM_010716.3 | NP_034846.2 | 13, 14 | 2166 | |||
GenBank mRNA | AK031540.1 | - | 13, 14 | 2103 | ||
AK143531.1 | - | 13, 14 | 2032 | |||
AK150644.1 | - | 13, 14 | 2071 | |||
AK156018.1 | - | 13, 14 | 2081 | |||
AK156030.1 | - | 2, 3 | 148 | |||
AK156825.1 | - | 2, 3 | 148 | |||
BC026143.1 | - | 12, 13 | 2390 | |||
BC049240.1 | - | 13, 14 | 2069 | 4504384 | ||
BC083500.1 | - | 13, 14 | 2065 | 3673103 | ||
U66057.1 | - | 13, 14 | 2097 | |||
U66058.1 | - | 13, 14 | 2097 |