Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291245.1 | NP_001278174.1 | 4 | 933 | ||
NM_001291246.1 | NP_001278175.1 | 4 | 930 | |||
NM_001291247.1 | NP_001278176.1 | 4 | 934 | |||
NM_010716.3 | NP_034846.2 | 4 | 937 | |||
GenBank mRNA | AK031540.1 | - | 4 | 875 | ||
AK143531.1 | - | 4 | 803 | |||
AK150644.1 | - | 4 | 842 | |||
AK156018.1 | - | 4 | 852 | |||
AK167363.1 | - | 4 | 845 | |||
BC026143.1 | - | 4 | 905 | |||
BC049240.1 | - | 4 | 840 | 4504384 | ||
BC083500.1 | - | 4 | 836 | 3673103 | ||
U66057.1 | - | 4 | 862 | |||
U66058.1 | - | 4 | 862 |