Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141813.1 | - | 6 | 849 | ||
AK164484.1 | - | 6 | 851 | |||
AK170166.1 | - | 5 | 624 | |||
AK172083.1 | - | 5 | 608 | |||
BC063768.1 | - | 6 | 831 | |||
CV563459.1 | - | 5 | 608 | |||
U14166.1 | - | 6 | 872 | |||
U15159.1 | - | 6 | 847 |