Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034030.2 | NP_001029202.1 | 9 | 1024 | ||
NM_010718.4 | NP_034848.1 | 13 | 1663 | |||
NM_173053.1 | NP_774958.1 | 12 | 1794 | |||
GenBank mRNA | AB005131.1 | - | 13 | 1516 | ||
AB005132.1 | - | 12 | 1778 | |||
AB008117.1 | - | 13 | 1599 | |||
AB012092.1 | - | 9 | 1024 | |||
AB012291.1 | - | 9 | 1024 | |||
AK034900.1 | - | 4 | 325 | |||
AK146099.1 | - | 12 | 1751 | |||
AK189591.1 | - | 2 | 141 | |||
BC007129.1 | - | 13 | 1606 | 3496836 | ||
BC086629.1 | - | 12 | 1724 | |||
U88618.1 | - | 13 | 1516 |