Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111100.1 | NP_001104570.1 | 6 | 717 | ||
NM_021460.3 | NP_067435.3 | 6 | 721 | |||
GenBank mRNA | AK088659.1 | - | 6 | 673 | ||
AK149791.1 | - | 6 | 681 | |||
AK154804.1 | - | 6 | 663 | |||
AK164007.1 | - | 6 | 717 | |||
AK169567.1 | - | 6 | 687 | |||
AK187548.1 | - | 3 | 289 | |||
AK218131.1 | - | 3 | 289 | |||
BC058564.1 | - | 6 | 669 | 6441278 | ||
Z31689.1 | - | 5 | 626 |