Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010721.2 | NP_034851.2 | 3 | 921 | ||
GenBank mRNA | AK079511.1 | - | 3 | 922 | ||
AK089110.1 | - | 3 | 888 | |||
AK132252.1 | - | 3 | 923 | |||
BC052729.1 | - | 3 | 870 | 6816118 | ||
BC058392.1 | - | 3 | 870 | 6816118 | ||
M35153.1 | - | 3 | 876 | |||
X16705.1 | - | 3 | 875 |