Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028796.1 | - | 9 | 1769 | ||
AK156499.1 | - | 7 | 1728 | |||
AK157297.1 | - | 8 | 1638 | |||
AK165124.1 | - | 6 | 958 | |||
AK171128.1 | - | 9 | 1712 | |||
AK172551.1 | - | 8 | 1606 | |||
AK172573.1 | - | 8 | 1642 | |||
AK185577.1 | - | 2 | 62 | |||
AK204703.1 | - | 2 | 62 | |||
BC006759.1 | - | 8 | 1622 | 3709861 | ||
U89992.2 | - | 1533 |