Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000806.5 | NP_000797.2 | 8 | 1110 | ||
NM_001127643.1 | NP_001121115.1 | 8 | 997 | |||
NM_001127644.1 | NP_001121116.1 | 7 | 1146 | |||
NM_001127645.1 | NP_001121117.1 | 7 | 734 | |||
NM_001127648.1 | NP_001121120.1 | 6 | 756 | |||
GenBank mRNA | AK223447.1 | - | 7 | 883 | ||
AK289792.1 | - | 8 | 1110 | |||
AK295160.1 | - | 8 | 929 | |||
BC030696.1 | - | 7 | 711 | 4820972 | ||
DA232066.1 | - | 3 | 406 | |||
X14766.1 | - | 7 | 856 |