Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010731.3 | NP_034861.3 | 3 | 1849 | ||
GenBank mRNA | AF086830.1 | - | 3 | 1466 | ||
AK010379.1 | - | 2 | 337 | |||
AK149961.1 | - | 3 | 1527 | |||
AK154907.1 | - | 3 | 1543 | |||
BC024815.2 | - | 3 | 1849 | |||
BC057204.1 | - | 3 | 1487 | |||
BC138524.1 | - | 3 | 1608 | 8861544 | ||
BC145311.1 | - | 3 | 1648 | 9053866 | ||
BC171957.1 | - | 3 | 1608 | 9054243 |