Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271708.1 | NP_001258637.1 | 3 | 1028 | ||
NM_001271709.1 | NP_001258638.1 | 2 | 994 | |||
NM_010733.3 | NP_034863.1 | 2 | 999 | |||
GenBank mRNA | AK036316.1 | - | 2 | 897 | ||
BC069041.1 | - | 2 | 693 | 5698788 | ||
D49802.1 | - | 2 | 910 |