Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010734.2 | NP_034864.2 | 4 | 262 | ||
GenBank mRNA | AF000428.1 | - | 4 | 213 | ||
AI182980.1 | - | 4 | 261 | |||
AI323110.1 | - | 3 | 167 | |||
AK146409.1 | - | 3 | 120 | |||
BC086881.1 | - | 4 | 256 | 6747449 | ||
M18187.1 | - | 4 | 254 | |||
U72643.1 | - | 4 | 250 |