Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008522.3 | NP_032548.2 | 7 | 803 | ||
GenBank mRNA | AK036491.1 | - | 7 | 804 | ||
AK144556.1 | - | 7 | 804 | |||
AK150799.1 | - | 7 | 804 | |||
AK151822.1 | - | 7 | 804 | |||
AK164781.1 | - | 7 | 805 | |||
AY792500.1 | - | 7 | 769 | |||
BC006904.1 | - | 7 | 811 | 3600033 | ||
BC008530.1 | - | 7 | 784 | |||
BC009662.1 | - | 3 | 234 | |||
CT010339.1 | - | 7 | 769 | |||
D88510.1 | - | 7 | 803 | |||
FJ538998.1 | - | 7 | 769 | |||
J03298.1 | - | 7 | 772 |