Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000413.2 | NP_000404.2 | 1, 2 | 1059 | ||
XM_011524729.1 | XP_011523031.1 | 1, 2 | 467 | |||
XM_011524730.1 | XP_011523032.1 | 1, 2 | 467 | |||
XM_011524731.1 | XP_011523033.1 | 1, 2 | 467 | |||
GenBank mRNA | AK127832.1 | - | 1, 2 | 3811 | ||
AK300143.1 | - | 1, 2 | 102 | |||
AK300159.1 | - | 1, 2 | 104 | |||
AK300452.1 | - | 1, 2 | 102 | |||
AK316292.1 | - | 1, 2 | 287 | |||
BC104752.1 | - | 1, 2 | 117 | 8143755 | ||
BC111935.1 | - | 1, 2 | 117 | 8327403 | ||
M36263.1 | - | 1, 2 | 100 | |||
X13440.1 | - | 1, 2 | 100 |