Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159902.1 | NP_001153374.1 | 3 | 379 | ||
NM_010737.3 | NP_034867.3 | 4 | 478 | |||
GenBank mRNA | AF354262.1 | - | 4 | 377 | ||
BC120707.1 | - | 4 | 416 | 40129630 | ||
BC145045.1 | - | 3 | 317 | 9053593 | ||
DQ237927.1 | - | 4 | 364 | |||
HQ713432.1 | - | 4 | 359 | |||
M77676.1 | - | 4 | 478 | |||
M77753.1 | - | 4 | 377 | |||
X64723.1 | - | 4 | 335 |