Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008532.2 | NP_032558.2 | 6 | 980 | ||
GenBank mRNA | AK133510.1 | - | 5 | 662 | ||
AK145752.1 | - | 5 | 556 | |||
AK167182.1 | - | 5 | 625 | |||
BC005618.1 | - | 5 | 621 | 3711069 | ||
BC094465.1 | - | 5 | 646 | 4948935 | ||
M76124.1 | - | 5 | 629 |