Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010746.3 | NP_034876.2 | 3, 4 | 378 | ||
GenBank mRNA | AJ223765.1 | - | 3, 4 | 367 | ||
AK079401.1 | - | 3, 4 | 379 | |||
AK172251.1 | - | 2, 3 | 411 | |||
BC042788.1 | - | 3, 4 | 348 | |||
BC115364.2 | - | 3, 4 | 379 | 40045898 |