Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008535.2 | NP_032561.2 | 4 | 1773 | ||
GenBank mRNA | AK076114.1 | - | 4 | 1426 | ||
BC005736.1 | - | 4 | 1392 | 3598911 |