Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008535.2 | NP_032561.2 | 2 | 608 | ||
GenBank mRNA | AK076114.1 | - | 2 | 262 | ||
BC005736.1 | - | 2 | 228 | 3598911 | ||
CT010167.1 | - | 1 | 53 | |||
X57687.1 | - | 2 | 518 |