Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010749.6 | NP_034879.2 | 5 | 734 | ||
GenBank mRNA | AK004656.1 | - | 4 | 2377 | ||
AK133943.1 | - | 5 | 740 | |||
AK145913.1 | - | 5 | 746 | |||
AK149177.1 | - | 5 | 764 | |||
AK151607.1 | - | 5 | 795 | |||
AK153140.1 | - | 5 | 746 | |||
AK153302.1 | - | 5 | 815 | |||
AK153474.1 | - | 5 | 755 | |||
AK196390.1 | - | 1 | 122 | |||
AK197278.1 | - | 1 | 122 | |||
BC027210.1 | - | 5 | 689 | |||
BC046956.1 | - | 5 | 734 | |||
BC080811.1 | - | 5 | 734 | 6438831 | ||
M58585.1 | - | 1 | 112 | |||
M63286.1 | - | 5 | 642 | |||
X56831.1 | - | 5 | 674 | |||
X64068.1 | - | 5 | 737 |
Gene Symbol: M6pr-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X64069.1 | - | 1 | 1100 |