Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010749.6 | NP_034879.2 | 3, 4 | 499 | ||
GenBank mRNA | AK133943.1 | - | 3, 4 | 505 | ||
AK145913.1 | - | 3, 4 | 511 | |||
AK149177.1 | - | 3, 4 | 529 | |||
AK151607.1 | - | 3, 4 | 560 | |||
AK153140.1 | - | 3, 4 | 511 | |||
AK153302.1 | - | 3, 4 | 580 | |||
AK153474.1 | - | 3, 4 | 520 | |||
BC027210.1 | - | 3, 4 | 454 | |||
BC046956.1 | - | 3, 4 | 499 | |||
BC080811.1 | - | 3, 4 | 499 | 6438831 | ||
M63286.1 | - | 3, 4 | 407 | |||
X56831.1 | - | 3, 4 | 439 | |||
X64068.1 | - | 3, 4 | 502 |