Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010749.6 | NP_034879.2 | 6 | 766 | ||
GenBank mRNA | AK004656.1 | - | 5 | 2409 | ||
AK133943.1 | - | 6 | 772 | |||
AK145913.1 | - | 6 | 778 | |||
AK151607.1 | - | 6 | 827 | |||
AK153302.1 | - | 6 | 847 | |||
AK153474.1 | - | 6 | 787 | |||
BC027210.1 | - | 6 | 721 | |||
BC046956.1 | - | 6 | 766 | |||
BC080811.1 | - | 6 | 766 | 6438831 | ||
M58585.1 | - | 2 | 144 | |||
M63286.1 | - | 6 | 674 | |||
X56831.1 | - | 6 | 706 | |||
X64068.1 | - | 6 | 769 |
Gene Symbol: M6pr-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X64069.1 | - | 1 | 1132 |