Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010752.3 | NP_034882.2 | 10 | 1074 | ||
GenBank mRNA | AF083812.1 | - | 10 | 1081 | ||
AF083813.1 | - | 10 | 1081 | |||
AK040246.1 | - | 2 | 109 | |||
AK136981.1 | - | 10 | 1077 | |||
BC010702.1 | - | 10 | 1072 | 3156639 |