Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010752.3 | NP_034882.2 | 2 | 136 | ||
GenBank mRNA | AF083812.1 | - | 2 | 143 | ||
AF083813.1 | - | 2 | 143 | |||
AK136981.1 | - | 2 | 138 | |||
AK200567.1 | - | 2 | 122 | |||
BC010702.1 | - | 2 | 134 | 3156639 | ||
BI735882.1 | - | 2 | 136 |