Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005526.2 | NP_005517.1 | 9, 10 | 1315 | ||
XM_017013377.1 | XP_016868866.1 | 9, 10 | 1315 | |||
GenBank mRNA | AF299094.1 | - | 1, 2 | 35 | ||
AK054762.1 | - | 5, 6 | 1432 | |||
AK125467.1 | - | 6, 7 | 2617 | |||
AK222497.1 | - | 9, 10 | 1299 | |||
AK290975.1 | - | 9, 10 | 1295 | |||
AK300205.1 | - | 6, 7 | 797 | |||
BC014638.1 | - | 9, 10 | 1286 | 3528599 | ||
BT007351.1 | - | 9, 10 | 1145 | |||
M64673.1 | - | 9, 10 | 1305 |