Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005526.2 | NP_005517.1 | 2 | 306 | ||
XM_005272315.2 | XP_005272372.1 | 2 | 306 | |||
XM_005272316.2 | XP_005272373.1 | 2 | 306 | |||
XM_005272317.1 | XP_005272374.1 | 2 | 306 | |||
XM_017013377.1 | XP_016868866.1 | 2 | 306 | |||
GenBank mRNA | AK222497.1 | - | 2 | 290 | ||
AK290975.1 | - | 2 | 286 | |||
BC014638.1 | - | 2 | 277 | 3528599 | ||
BT007351.1 | - | 2 | 136 | |||
DA481620.1 | - | 2 | 306 | |||
M64673.1 | - | 2 | 296 |