Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145835.1 | - | 3 | 1631 | ||
AK146043.1 | - | 3 | 1600 | |||
AK156896.1 | - | 3 | 1662 | |||
BC022952.1 | - | 3 | 1522 | 5132212 | ||
BX515677.1 | - | 1 | 244 |