Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magea8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020020.4 | NP_064404.1 | 4 | 1448 | ||
GenBank mRNA | AK145480.1 | - | 4 | 1449 |
Gene Symbol: Magea5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020018.1 | NP_064402.1 | 3 | 1364 |