Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282737.1 | NP_001269666.1 | 3 | 267 | ||
GenBank mRNA | AF007862.1 | - | 3 | 254 | ||
AF035939.1 | - | 3 | 214 | |||
AK011512.1 | - | 3 | 206 | |||
BC018176.1 | - | 3 | 205 | 3587774 | ||
BY134232.1 | - | 3 | 268 |