Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccndbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010761.2 | NP_034891.2 | 6 | 562 | ||
GenBank mRNA | AK144883.1 | - | 6 | 539 | ||
AK160205.1 | - | 6 | 528 | |||
AK163150.1 | - | 6 | 475 | |||
AK167345.1 | - | 6 | 527 | |||
AK186314.1 | - | 5 | 339 | |||
CO810054.1 | - | 6 | 569 | |||
U50734.1 | - | 6 | 480 |