Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008548.4 | NP_032574.1 | 8 | 1688 | ||
GenBank mRNA | AK031038.1 | - | 8 | 1689 | ||
AK042371.1 | - | 7 | 1529 | |||
AK043932.1 | - | 8 | 1629 | |||
AK154820.1 | - | 8 | 1630 | |||
AK171841.1 | - | 8 | 1631 | |||
BC015265.1 | - | 8 | 1589 | 4223319 | ||
U04299.1 | - | 7 | 1292 |