Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008548.4 | NP_032574.1 | 10 | 1864 | ||
GenBank mRNA | AK031038.1 | - | 10 | 1865 | ||
AK042371.1 | - | 9 | 1705 | |||
AK043932.1 | - | 10 | 1805 | |||
AK154820.1 | - | 10 | 1806 | |||
AK171841.1 | - | 10 | 1807 | |||
BC015265.1 | - | 10 | 1765 | 4223319 | ||
U04299.1 | - | 9 | 1468 |