Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040667.2 | NP_001035757.1 | 7, 8 | 1534 | ||
NM_001538.3 | NP_001529.2 | 7, 8 | 1534 | |||
GenBank mRNA | AB029348.1 | - | 5, 6 | 576 | ||
AK225152.1 | - | 5, 6 | 591 | |||
AK293631.1 | - | 3, 4 | 1037 | |||
AK296210.1 | - | 5, 6 | 591 | |||
BC130383.1 | - | 5, 6 | 637 | |||
D87673.1 | - | 5, 6 | 576 |