Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008553.4 | NP_032579.2 | 1 | 837 | ||
GenBank mRNA | AK141866.1 | - | 1 | 827 | ||
AK143210.1 | - | 1 | 838 | |||
BC055748.1 | - | 1 | 359 | 6415061 | ||
JX478273.1 | - | 1 | 265 | |||
M95603.1 | - | 1 | 340 |