Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Masp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003893.2 | NP_001003893.1 | 2 | 260 | ||
NM_010767.3 | NP_034897.1 | 2 | 260 | |||
GenBank mRNA | AA238260.1 | - | 2 | 248 | ||
AB009459.1 | - | 2 | 250 | |||
AI529228.1 | - | 2 | 261 | |||
AJ250369.1 | - | 2 | 249 | |||
AK050052.1 | - | 2 | 237 | |||
AK149421.1 | - | 1 | 314 | |||
BC013893.1 | - | 1 | 234 | 4211293 | ||
BF233497.1 | - | 2 | 258 | |||
Y19160.1 | - | 1 | 307 | |||
Y19163.1 | - | 1 | 277 |