Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285853.1 | NP_001272782.1 | 8 | 1006 | ||
NM_001285854.1 | NP_001272783.1 | 8 | 1003 | |||
NM_001285855.1 | NP_001272784.1 | 8 | 1071 | |||
NM_010768.2 | NP_034898.1 | 9 | 1139 | |||
GenBank mRNA | AK013606.1 | - | 8 | 820 | ||
AK079816.1 | - | 7 | 705 | |||
AK158883.1 | - | 7 | 770 | |||
BC060086.1 | - | 6 | 1967 | |||
BC111864.1 | - | 8 | 817 | 40046807 | ||
D45243.1 | - | 8 | 1071 | |||
L27738.1 | - | 9 | 893 | |||
S77473.1 | - | 8 | 818 | |||
U05210.1 | - | 8 | 792 |