Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010773.2 | NP_034903.2 | 3 | 906 | ||
GenBank mRNA | AF072243.1 | - | 3 | 864 | ||
AK035891.1 | - | 1 | 1385 | |||
AK147008.1 | - | 3 | 791 | |||
BC026698.1 | - | 3 | 866 | |||
BC046607.1 | - | 3 | 906 | 6399206 | ||
BC057289.1 | - | 3 | 738 | |||
BC058864.1 | - | 3 | 726 |