Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005345.5 | NP_005336.3 | 1 | 2218 | ||
GenBank mRNA | AB209818.1 | - | 1 | 2179 | ||
AK096017.1 | - | 2 | 1694 | |||
AK225692.1 | - | 1 | 2189 | |||
AK225699.1 | - | 1 | 2189 | |||
AK291295.1 | - | 1 | 2189 | |||
AK294182.1 | - | 2 | 1982 | |||
AK298056.1 | - | 2 | 2117 | |||
AK298098.1 | - | 2 | 1517 | |||
AK301243.1 | - | 2 | 1898 | |||
AK301574.1 | - | 2 | 2135 | |||
AK303980.1 | - | 2 | 1460 | |||
AK304618.1 | - | 2 | 1955 | |||
AK304652.1 | - | 2 | 2024 | |||
BC002453.2 | - | 1 | 2152 | 3345864 | ||
FJ224292.1 | - | 1 | 2464 |