Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010774.2 | NP_034904.2 | 7 | 1688 | ||
GenBank mRNA | AF072249.1 | - | 7 | 1705 | ||
AK147782.1 | - | 4 | 670 | |||
AK161005.1 | - | 7 | 2676 | |||
BC024812.1 | - | 7 | 1670 | 5356016 |