Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178076.3 | NP_835177.2 | 27 | 3075 | ||
GenBank mRNA | AK147252.1 | - | 24 | 3279 | ||
BC058622.1 | - | 27 | 3010 | 5704822 |