Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033196.1 | - | 16 | 2085 | ||
AK051795.1 | - | 16 | 2055 | |||
AK088558.1 | - | 9 | 1001 | |||
AK133826.1 | - | 16 | 2086 | |||
BC090645.1 | - | 16 | 2038 | |||
BC094416.1 | - | 16 | 2050 | 5707724 | ||
D26090.1 | - | 15 | 2105 |