Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008568.2 | NP_032594.1 | 2 | 166 | ||
GenBank mRNA | AK132748.1 | - | 727 | |||
AK133091.1 | - | 2 | 162 | |||
AK144980.1 | - | 2 | 178 | |||
AK149913.1 | - | 2 | 163 | |||
AK150059.1 | - | 2 | 163 | |||
AK150066.1 | - | 2 | 163 | |||
AK154052.1 | - | 2 | 163 | |||
BC023232.1 | - | 2 | 142 | |||
BC065164.1 | - | 2 | 150 | 6829737 | ||
BC066024.1 | - | 2 | 105 | 6829337 | ||
D26091.1 | - | 2 | 166 |