Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012235.3 | NP_036367.2 | 6 | 950 | ||
XM_005264967.1 | XP_005265024.1 | 6 | 877 | |||
XM_011533501.1 | XP_011531803.1 | 6 | 977 | |||
XM_017005918.1 | XP_016861407.1 | 6 | 964 | |||
XM_017005919.1 | XP_016861408.1 | 6 | 897 | |||
XM_017005920.1 | XP_016861409.1 | 6 | 897 | |||
GenBank mRNA | BC027207.2 | - | 2 | 149 | ||
D83782.2 | - | 6 | 936 |