Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpsab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031187.4 | NP_112464.4 | 3 | 587 | ||
GenBank mRNA | AK040500.1 | - | 2 | 422 | ||
BC011328.1 | - | 3 | 552 | 3994444 | ||
L00653.1 | - | 3 | 418 | |||
U42405.1 | - | 3 | 306 |